Assay Details
| Gene Symbol: LCAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111202.4 | NP_566201.1 | 6 - 7 | 947 | 106 | |
| GenBank mRNA | AF421148.1 | - | 6 - 7 | 732 | 106 | |
| AY054239.1 | - | 6 - 7 | 765 | 106 | ||
| AY056414.1 | - | 6 - 7 | 822 | 106 | ||
| AY113174.1 | - | 6 - 7 | 732 | 106 | ||
More Information
Set Membership: |
|

LCAT3
Genomic Map