Assay Details
| Gene Symbol: AT3G03440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111215.4 | NP_186994.2 | 1 - 2 | 572 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G03440| Gene Symbol: AT3G03440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111215.4 | NP_186994.2 | 1 - 2 | 572 | 128 | |
Set Membership: |
|