Assay Details
| Gene Symbol: NPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111225.2 | NP_566207.1 | 1 - 2 | 446 | 83 | |
| GenBank mRNA | AB084294.1 | - | 1 - 2 | 446 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPC5| Gene Symbol: NPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111225.2 | NP_566207.1 | 1 - 2 | 446 | 83 | |
| GenBank mRNA | AB084294.1 | - | 1 - 2 | 446 | 83 | |
Set Membership: |
|