Assay Details
| Gene Symbol: CS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111234.5 | NP_187013.1 | 1 - 2 | 370 | 68 | |
| GenBank mRNA | AB003041.1 | - | 1 - 2 | 349 | 68 | |
| AY099573.1 | - | 1 - 2 | 355 | 68 | ||
| BT002155.1 | - | 1 - 2 | 309 | 68 | ||
More Information
Set Membership: |
|

CS26
Genomic Map