Assay Details
| Gene Symbol: CS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111234.5 | NP_187013.1 | 5 - 6 | 948 | 101 | |
| GenBank mRNA | AB003041.1 | - | 5 - 6 | 927 | 101 | |
| AY099573.1 | - | 5 - 6 | 933 | 101 | ||
| BT002155.1 | - | 5 - 6 | 887 | 101 | ||
More Information
Set Membership: |
|

CS26
Genomic Map