Assay Details
| Gene Symbol: CS26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111234.5 | NP_187013.1 | 7 - 8 | 1049 | 87 | |
| GenBank mRNA | AB003041.1 | - | 7 - 8 | 1028 | 87 | |
| AY099573.1 | - | 7 - 8 | 1034 | 87 | ||
| BT002155.1 | - | 7 - 8 | 988 | 87 | ||
More Information
Set Membership: |
|

CS26
Genomic Map