Assay Details
| Gene Symbol: BGLU25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111235.4 | NP_187014.1 | 11 - 12 | 1305 | 120 | |
| GenBank mRNA | AF082157.1 | - | 11 - 12 | 1269 | 120 | |
More Information
Set Membership: |
|

BGLU25
Genomic Map