Assay Details
| Gene Symbol: BGLU25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111235.4 | NP_187014.1 | 6 - 7 | 570 | 96 | |
| GenBank mRNA | AF082157.1 | - | 6 - 7 | 534 | 96 | |
More Information
Set Membership: |
|

BGLU25
Genomic Map