Assay Details
| Gene Symbol: CAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161114.2 | NP_001154586.1 | 10 - 11 | 1654 | 140 | |
| GenBank mRNA | AY062508.1 | - | 10 - 11 | 1567 | 140 | |
| BT008369.1 | - | 10 - 11 | 1352 | 140 | ||
More Information
Set Membership: |
|

CAT4
Genomic Map