Assay Details
| Gene Symbol: TSF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118156.2 | NP_193770.1 | 3 - 4 | 344 | 79 | |
| GenBank mRNA | AB027506.1 | - | 3 - 4 | 330 | 79 | |
| AF152907.1 | - | 3 - 4 | 299 | 79 | ||
More Information
Set Membership: |
|

TSF
Genomic Map