Assay Details
| Gene Symbol: AT4G20390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118158.4 | NP_193772.1 | 2 - 3 | 594 | 63 | |
| GenBank mRNA | BT025759.1 | - | 2 - 3 | 341 | 63 | |
More Information
Set Membership: |
|

AT4G20390
Genomic Map