Assay Details
| Gene Symbol: smB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036605.1 | NP_001031682.1 | 2 | 1350 | 134 | |
| NM_001341439.1 | NP_001329619.1 | 2 | 1076 | 134 | ||
| NM_118163.4 | NP_193777.1 | 1 | 1651 | 134 | ||
| NM_179083.3 | NP_849414.1 | 2 | 1547 | 134 | ||
| GenBank mRNA | AK317021.1 | - | 2 | 1347 | 134 | |
More Information
Set Membership: |
|

smB
Genomic Map