Assay Details
| Gene Symbol: AAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118231.4 | NP_193844.2 | 1 - 2 | 748 | 64 | |
| GenBank mRNA | AK227519.1 | - | 1 - 2 | 688 | 64 | |
| BT006445.1 | - | 1 - 2 | 415 | 64 | ||
More Information
Set Membership: |
|

AAT1
Genomic Map