Assay Details
| Gene Symbol: HAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203855.2 | NP_001190784.2 | 9 - 10 | 917 | 83 | |
| NM_001203856.1 | NP_001190785.1 | 10 - 11 | 1068 | 83 | ||
| NM_118234.4 | NP_193847.2 | 10 - 11 | 1029 | 83 | ||
| GenBank mRNA | AY058845.1 | - | 10 - 11 | 1029 | 83 | |
| AY079034.1 | - | 9 - 10 | 917 | 83 | ||
More Information
Set Membership: |
|

HAP6
Genomic Map