Assay Details
| Gene Symbol: HSA32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118250.5 | NP_567623.2 | 2 - 3 | 446 | 63 | |
| GenBank mRNA | AK118775.1 | - | 2 - 3 | 359 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSA32| Gene Symbol: HSA32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118250.5 | NP_567623.2 | 2 - 3 | 446 | 63 | |
| GenBank mRNA | AK118775.1 | - | 2 - 3 | 359 | 63 | |
Set Membership: |
|