Assay Details
| Gene Symbol: RK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341478.1 | NP_001328871.1 | 3 - 4 | 1767 | 99 | |
| NM_118258.3 | NP_193869.1 | 3 - 4 | 1767 | 99 | ||
| GenBank mRNA | AK118895.1 | - | 3 - 4 | 1767 | 99 | |
| BT006032.1 | - | 3 - 4 | 1631 | 99 | ||
More Information
Set Membership: |
|

RK3
Genomic Map