Assay Details
| Gene Symbol: RK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118258.3 | NP_193869.1 | 4 - 5 | 1972 | 118 | |
| GenBank mRNA | AK118895.1 | - | 4 - 5 | 1972 | 118 | |
| BT006032.1 | - | 4 - 5 | 1836 | 118 | ||
More Information
Set Membership: |
|

RK3
Genomic Map