Assay Details
| Gene Symbol: RK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341478.1 | NP_001328871.1 | 5 - 6 | 2452 | 65 | |
| NM_118258.3 | NP_193869.1 | 6 - 7 | 2361 | 65 | ||
| GenBank mRNA | AK118895.1 | - | 6 - 7 | 2361 | 65 | |
| BT006032.1 | - | 6 - 7 | 2225 | 65 | ||
More Information
Set Membership: |
|

RK3
Genomic Map