Assay Details
| Gene Symbol: CRK28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341480.1 | NP_001320019.1 | 2 - 3 | 1008 | 140 | |
| NM_001341483.1 | NP_001328836.1 | 2 - 3 | 1008 | 140 | ||
| NM_001341484.1 | NP_001328837.1 | 2 - 3 | 1008 | 140 | ||
| NM_001341485.1 | NP_001328838.1 | 2 - 3 | 1008 | 140 | ||
More Information
Set Membership: |
|

CRK28
Genomic Map