Assay Details
| Gene Symbol: SPHK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036612.2 | NP_001031689.2 | 1 - 2 | 402 | 72 | |
| NM_001341496.1 | NP_001320022.1 | 1 - 2 | 727 | 72 | ||
| NM_118274.8 | NP_193885.6 | 1 - 2 | 727 | 72 | ||
| GenBank mRNA | AY128394.1 | - | 1 - 2 | 395 | 72 | |
| BT006621.1 | - | 1 - 2 | 346 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001036612.2 transcripts.
Set Membership: |
|

SPHK1
Genomic Map