Assay Details
| Gene Symbol: VAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341497.1 | NP_001320023.1 | 7 - 8 | 1433 | 152 | |
| NM_001341498.1 | NP_001329314.1 | 7 - 8 | 1392 | 152 | ||
| NM_118275.4 | NP_193886.3 | 7 - 8 | 1373 | 152 | ||
More Information
Set Membership: |
|

VAL3
Genomic Map