Assay Details
| Gene Symbol: BGLU47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341512.1 | NP_001328261.1 | 4 - 5 | 526 | 120 | |
| NM_118296.4 | NP_193907.2 | 4 - 5 | 448 | 120 | ||
More Information
Set Membership: |
|

BGLU47
Genomic Map