Assay Details
| Gene Symbol: MSRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084954.2 | NP_001078423.1 | 1 - 2 | 363 | 96 | |
| NM_001203862.1 | NP_001190791.1 | 1 - 2 | 363 | 96 | ||
| NM_001341518.1 | NP_001320026.1 | 1 - 2 | 363 | 96 | ||
| NM_118306.7 | NP_567639.1 | 1 - 2 | 510 | 96 | ||
| GenBank mRNA | AF360341.1 | - | 1 - 2 | 346 | 96 | |
| AY051078.1 | - | 1 - 2 | 293 | 96 | ||
More Information
Set Membership: |
|

MSRB2
Genomic Map