Assay Details
| Gene Symbol: SRF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125558.1 | NP_001119030.1 | 9 - 10 | 1456 | 66 | |
| NM_118334.5 | NP_193944.2 | 13 - 14 | 2149 | 66 | ||
| GenBank mRNA | AY518293.1 | - | 13 - 14 | 1912 | 66 | |
| FJ708751.1 | - | 13 - 14 | 1804 | 66 | ||
More Information
Set Membership: |
|

SRF8
Genomic Map