Assay Details
| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036620.2 | NP_001031697.1 | 6 - 7 | 1170 | 71 | |
| NM_118360.3 | NP_193965.1 | 7 - 8 | 1062 | 71 | ||
| NM_202862.2 | NP_974591.1 | 7 - 8 | 907 | 71 | ||
| GenBank mRNA | AY065310.1 | - | 7 - 8 | 985 | 71 | |
| AY084570.1 | - | 7 - 8 | 969 | 71 | ||
| AY091315.1 | - | 6 - 7 | 830 | 71 | ||
More Information
Set Membership: |
|

CDS2
Genomic Map