Assay Details
| Gene Symbol: AT4G22460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118372.2 | NP_193977.1 | 1 | 236 | 80 | |
| Gene Symbol: AT4G22463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341544.1 | NP_001329812.1 | 2 | 393 | 80 | |
More Information
Set Membership: |
|

AT4G22460
Genomic Map