Assay Details
| Gene Symbol: AT4G22490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118375.3 | NP_193980.1 | 1 | 564 | 89 | |
| GenBank mRNA | AY086721.1 | - | 1 | 458 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G22490| Gene Symbol: AT4G22490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118375.3 | NP_193980.1 | 1 | 564 | 89 | |
| GenBank mRNA | AY086721.1 | - | 1 | 458 | 89 | |
Set Membership: |
|