Assay Details
| Gene Symbol: AT4G22520 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118378.1 | NP_193983.1 | 1 | 318 | 107 | |
| Gene Symbol: AT4G22530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118379.5 | NP_193984.1 | 2 | 1399 | 107 | |
More Information
Set Membership: |
|

AT4G22520
Genomic Map