Assay Details
Gene Symbol: AT4G22790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118408.2 | NP_194010.1 | 1 | 1525 | 128 | |
GenBank mRNA | BT002796.1 | - | 1 | 1525 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
