Assay Details
| Gene Symbol: PGR5-LIKE A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084962.1 | NP_001078431.1 | 8 - 9 | 1073 | 65 | |
| NM_001084963.1 | NP_001078432.1 | 8 - 9 | 1076 | 65 | ||
| NM_118418.4 | NP_567672.1 | 8 - 9 | 1082 | 65 | ||
| NM_179092.1 | NP_849423.1 | 8 - 9 | 1082 | 65 | ||
| GenBank mRNA | BT000905.1 | - | 8 - 9 | 911 | 65 | |
More Information
Set Membership: |
|

PGR5-LIKE A
Genomic Map