Assay Details
| Gene Symbol: FZR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118420.5 | NP_194022.3 | 2 - 3 | 950 | 77 | |
| GenBank mRNA | AY099585.1 | - | 2 - 3 | 681 | 77 | |
| AY128834.1 | - | 2 - 3 | 615 | 77 | ||
More Information
Set Membership: |
|

FZR2
Genomic Map