Assay Details
| Gene Symbol: AGL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341568.1 | NP_001320037.1 | 3 - 4 | 608 | 150 | |
| NM_001341569.1 | NP_001328192.1 | 3 - 4 | 608 | 150 | ||
| NM_118424.2 | NP_194026.1 | 2 - 3 | 258 | 150 | ||
| GenBank mRNA | AF312664.1 | - | 2 - 3 | 258 | 150 | |
| AY142523.1 | - | 2 - 3 | 258 | 150 | ||
More Information
Set Membership: |
|

AGL19
Genomic Map