Assay Details
| Gene Symbol: AT4G22960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118425.3 | NP_194027.2 | 4 - 5 | 1058 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G22960| Gene Symbol: AT4G22960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118425.3 | NP_194027.2 | 4 - 5 | 1058 | 131 | |
Set Membership: |
|