Assay Details
| Gene Symbol: GSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036624.1 | NP_001031701.1 | 12 - 13 | 1422 | 116 | |
| NM_001203879.2 | NP_001190808.1 | 12 - 13 | 1818 | 116 | ||
| NM_118439.4 | NP_194041.1 | 12 - 13 | 1688 | 116 | ||
| GenBank mRNA | AF419576.1 | - | 12 - 13 | 1404 | 116 | |
| AY143970.1 | - | 11 - 12 | 1233 | 116 | ||
More Information
Set Membership: |
|

GSH1
Genomic Map