Assay Details
| Gene Symbol: GSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036624.1 | NP_001031701.1 | 9 - 10 | 1164 | 76 | |
| NM_001203879.2 | NP_001190808.1 | 9 - 10 | 1560 | 76 | ||
| NM_118439.4 | NP_194041.1 | 9 - 10 | 1430 | 76 | ||
| GenBank mRNA | AF419576.1 | - | 9 - 10 | 1146 | 76 | |
| AY143970.1 | - | 8 - 9 | 975 | 76 | ||
More Information
Set Membership: |
|

GSH1
Genomic Map