Assay Details
| Gene Symbol: CRK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341575.1 | NP_001329890.1 | 1 - 2 | 900 | 83 | |
| NM_118442.4 | NP_567677.1 | 1 - 2 | 894 | 83 | ||
| NM_179094.3 | NP_849425.1 | 1 - 2 | 894 | 83 | ||
| GenBank mRNA | AF224707.1 | - | 1 - 2 | 814 | 83 | |
| AY142496.1 | - | 1 - 2 | 814 | 83 | ||
More Information
Set Membership: |
|

CRK5
Genomic Map