Assay Details
| Gene Symbol: CRK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118444.3 | NP_194046.2 | 1 - 2 | 872 | 61 | |
| GenBank mRNA | AY133720.1 | - | 1 - 2 | 805 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRK7| Gene Symbol: CRK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118444.3 | NP_194046.2 | 1 - 2 | 872 | 61 | |
| GenBank mRNA | AY133720.1 | - | 1 - 2 | 805 | 61 | |
Set Membership: |
|