Assay Details
| Gene Symbol: CRK10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341579.1 | NP_001328631.1 | 4 - 5 | 1335 | 67 | |
| NM_001341580.1 | NP_001328630.1 | 4 - 5 | 1335 | 67 | ||
| NM_118447.3 | NP_567679.2 | 4 - 5 | 1477 | 67 | ||
| GenBank mRNA | AK117765.1 | - | 4 - 5 | 1335 | 67 | |
| BT005954.1 | - | 4 - 5 | 1301 | 67 | ||
More Information
Set Membership: |
|

CRK10
Genomic Map