Assay Details
| Gene Symbol: CRK24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341599.1 | NP_001320045.1 | 2 - 3 | 679 | 86 | |
| NM_001341600.1 | NP_001328061.1 | 1 - 2 | 772 | 86 | ||
| NM_001341601.1 | NP_001328060.1 | 2 - 3 | 630 | 86 | ||
More Information
Set Membership: |
|

CRK24
Genomic Map