Assay Details
| Gene Symbol: AT4G23530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118484.4 | NP_194084.1 | 1 | 1727 | 101 | |
| GenBank mRNA | AY063829.1 | - | 1 | 1653 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G23530| Gene Symbol: AT4G23530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118484.4 | NP_194084.1 | 1 | 1727 | 101 | |
| GenBank mRNA | AY063829.1 | - | 1 | 1653 | 101 | |
Set Membership: |
|