Assay Details
| Gene Symbol: AT4G23540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118485.9 | NP_194085.4 | 7 - 8 | 2092 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G23540| Gene Symbol: AT4G23540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118485.9 | NP_194085.4 | 7 - 8 | 2092 | 67 | |
Set Membership: |
|