Assay Details
| Gene Symbol: AT4G23590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118490.4 | NP_194090.2 | 1 - 2 | 319 | 98 | |
| GenBank mRNA | AY070389.1 | - | 1 - 2 | 319 | 98 | |
| AY123024.1 | - | 1 - 2 | 250 | 98 | ||
More Information
Set Membership: |
|

AT4G23590
Genomic Map