Assay Details
| Gene Symbol: AT2G03500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126400.3 | NP_027544.1 | 1 - 2 | 583 | 122 | |
| GenBank mRNA | AK221662.1 | - | 1 - 2 | 389 | 122 | |
| AY060559.1 | - | 1 - 2 | 389 | 122 | ||
| AY133630.1 | - | 1 - 2 | 202 | 122 | ||
More Information
Set Membership: |
|

AT2G03500
Genomic Map