Assay Details
| Gene Symbol: AT2G03500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126400.3 | NP_027544.1 | 2 - 3 | 1156 | 122 | |
| GenBank mRNA | AK221662.1 | - | 2 - 3 | 962 | 122 | |
| AY060559.1 | - | 2 - 3 | 962 | 122 | ||
| AY133630.1 | - | 2 - 3 | 775 | 122 | ||
More Information
Set Membership: |
|

AT2G03500
Genomic Map