Assay Details
| Gene Symbol: CDC48B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126415.6 | NP_178463.1 | 10 - 11 | 1385 | 118 | |
| GenBank mRNA | AY062443.1 | - | 10 - 11 | 1326 | 118 | |
| BT008371.1 | - | 10 - 11 | 1275 | 118 | ||
More Information
Set Membership: |
|

CDC48B
Genomic Map