Assay Details
| Gene Symbol: CDC48B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335198.1 | NP_001325037.1 | 4 - 5 | 523 | 134 | |
| NM_126415.6 | NP_178463.1 | 4 - 5 | 524 | 134 | ||
| GenBank mRNA | AY062443.1 | - | 4 - 5 | 465 | 134 | |
| BT008371.1 | - | 4 - 5 | 414 | 134 | ||
More Information
Set Membership: |
|

CDC48B
Genomic Map