Assay Details
| Gene Symbol: CDC48B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335198.1 | NP_001325037.1 | 6 - 7 | 722 | 85 | |
| NM_126415.6 | NP_178463.1 | 6 - 7 | 723 | 85 | ||
| GenBank mRNA | AY062443.1 | - | 6 - 7 | 664 | 85 | |
| BT008371.1 | - | 6 - 7 | 613 | 85 | ||
More Information
Set Membership: |
|

CDC48B
Genomic Map