Assay Details
| Gene Symbol: CDC48B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335198.1 | NP_001325037.1 | 7 - 8 | 1000 | 112 | |
| NM_126415.6 | NP_178463.1 | 7 - 8 | 1001 | 112 | ||
| GenBank mRNA | AY062443.1 | - | 7 - 8 | 942 | 112 | |
| BT008371.1 | - | 7 - 8 | 891 | 112 | ||
More Information
Set Membership: |
|

CDC48B
Genomic Map