Assay Details
| Gene Symbol: CDC48B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335198.1 | NP_001325037.1 | 9 - 10 | 1299 | 138 | |
| NM_126415.6 | NP_178463.1 | 9 - 10 | 1300 | 138 | ||
| GenBank mRNA | AY062443.1 | - | 9 - 10 | 1241 | 138 | |
| BT008371.1 | - | 9 - 10 | 1190 | 138 | ||
More Information
Set Membership: |
|

CDC48B
Genomic Map