Assay Details
| Gene Symbol: SPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161019.2 | NP_001154491.1 | 2 - 3 | 597 | 58 | |
| NM_001335199.1 | NP_001323957.1 | 2 - 3 | 620 | 58 | ||
| NM_126416.3 | NP_178464.1 | 2 - 3 | 396 | 58 | ||
| GenBank mRNA | AF412118.1 | - | 2 - 3 | 375 | 58 | |
| AY045933.1 | - | 2 - 3 | 379 | 58 | ||
| AY084847.1 | - | 2 - 3 | 383 | 58 | ||
| AY113867.1 | - | 1 - 2 | 289 | 58 | ||
| AY464947.1 | - | 2 - 3 | 370 | 58 | ||
More Information
Set Membership: |
|

SPR1
Genomic Map